About this item
CD31 (PECAM-1) is a transmembrane glycoprotein member of the immunoglobulin supergene family of adhesion molecules. CD31 is expressed by stem cells of the hematopoietic system and is primarily used to identify and concentrate these cells for experimental studies as well as for bone marrow transplantation. Anti-CD31 has shown to be highly specific and sensitive for vascular endothelial cells. Staining of nonvascular tumors (excluding hematopoietic neoplasms) is rare. CD31 MAb reacts with normal, benign, and malignant endothelial cells which make up blood vessel lining. The level of CD31 expression can help to determine the degree of tumor angiogenesis, and a high level of CD31 expression may imply a rapidly growing tumor and potentially a predictor of tumor recurrence.
Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone: C31.7
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Cynomolgus monkey, Human, Rabbit
Specifications
- Catalog No:
- BNC400014-500
- BNC400014-100
- Antigen symbol:
- PECAM1
- PECAM1
- Antigen name:
- Platelet Endothelial Cell Adhesion Molecule
- Platelet Endothelial Cell Adhesion Molecule
- Antigen synonyms:
- EndoCAM|GPIIA'|PECA1|Platelet Endothelial Cell Adhesion Molecule 1
- EndoCAM|GPIIA'|PECA1|Platelet Endothelial Cell Adhesion Molecule 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 640R
- CF® 640R
- Clone:
- Clone C31.7
- Clone C31.7
- Reactivity:
- Rabbit, Cynomolgus monkey, Human
- Rabbit, Cynomolgus monkey, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 376675 , 514412
- 376675 , 514412
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide