Anti-BCL2L1 Mouse Monoclonal Antibody (CF488A) [clone: BX006]
Supplier: Biotium
Certificates
About this item
Recognizes a protein of 27 kDa, identified as the Bcl-X protein. This MAb shows no cross-reaction with Bcl-2 or Bax protein. Bcl-X has two isoforms, Bcl-XL (long), a 241 amino acid protein which suppresses cell death. And Bcl-XS (short), a 178 amino acid protein lacking a 63 amino acid domain which functions as a dominant inhibitor of Bcl-2. This MAb reacts with both Bcl-XS and Bcl-XL proteins.
Type: Primary
Antigen: BCL2L1
Clonality: Monoclonal
Clone: BX006
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Pig, Rat
Specifications
- Catalog No:
- BNC880006-100
- BNC880006-500
- Antigen symbol:
- BCL2L1
- BCL2L1
- Antigen name:
- bcl-x
- bcl-x
- Antigen synonyms:
- BclXL|BCLX|Apoptosis regulator Bcl-X|BCL2L|Bcl2-L-1|Bcl-2-like protein 1|BCL XL/S|Protein phosphatase 1 regulatory subunit 52 (PPP1R52)|BclXs|B cell lymphoma 2 like|Bcl2l1
- BclXL|BCLX|Apoptosis regulator Bcl-X|BCL2L|Bcl2-L-1|Bcl-2-like protein 1|BCL XL/S|Protein phosphatase 1 regulatory subunit 52 (PPP1R52)|BclXs|B cell lymphoma 2 like|Bcl2l1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 488A
- CF® 488A
- Clone:
- Clone BX006
- Clone BX006
- Reactivity:
- Pig, Human, Rat, Mouse
- Pig, Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 516966
- 516966
- Isotype:
- IgG2a
- IgG2a
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Frequently Bought Together