Dulbecco's phosphate buffered saline (DPBS), cell culture, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Phosphate buffered salts, Dulbecco's formula (DPBS) is a balanced salt solution (BSS) used for the handling and culturing of mammalian cells. DPBS is used to to irrigate, wash, and dilute mammalian cells.
- Clear, colourless solution format
- pH: 7,0 to 7,6
- Stored at 15 to 30 °C
Phosphate buffering maintains the pH in the physiological range. Calcium and magnesium facilitate cell binding and clumping. DPBS without these ions can be used to wash and rinse suspended cells.
Frequently Bought Together