Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Indole-3-butyric acid potassium salt
Indole-3-butyric acid potassium salt ≥98%, powder
Indole-3-butyric acid potassium salt ≥98%, powder
ICNA0210204325
:
MP Biomedicals
:
Indole-3-butyric acid potassium salt ≥98%, powder
ICNA0210204325
:
MP Biomedicals
:
Pk:
25 g
Seq:
3308
Purity:
≥98%
FTIR:
Conforms to Standard
UV/VIS λmax (MeOH:
281 - 284 nm
:
:
241.331 g/mol
:
Refrigerator
:
60096-23-3
:
C₁₂H₁₂KNO₂
:
MFCD00058442