Bromophenol blue, powder
Catalog # ICNA04805732
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Identity Test:Passes
- UV/Vis: λmax (water):592 ± 4 nm (pH 7); 437 ± 3 nm (pH 3)
- Visual Transition Interval:Yellow (pH 3,0) to purple (pH 4,6)
Specification Test Results
About this item
:
:669.97 g/mol
Melting Pt:273 °C (lit.)
Storage Temperature:Ambient
CAS Number:115-39-9
Formula:C₁₉H₁₀Br₄O₅S
MDL Number:MFCD00005875