Methylene blue trihydrate, powder C.I. 52015
Catalog # ICNA04800663
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Identity Test:Passes
- UV/Vis: λmax (water):662 ± 3 nm
Specification Test Results
About this item
:
:373.9 g/mol
Storage Temperature:Ambient
CAS Number:7220-79-3
Formula:C₁₆H₁₈ClN₃S·3H₂O
MDL Number:MFCD00150008