Anti-NUMA1/D12 Mouse Monoclonal Antibody [clone: A73-B/D12]
Catalog # ABNOMAB13390
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:NUMA1/D12
- Clonality:Monoclonal
- Clone:A73-B/D12
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgM kappa
- Reactivity:Human
- Gene ID:4926
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against native human NUMA1.
Type: Primary
Antigen: NUMA1/D12
Clonality: Monoclonal
Clone: A73-B/D12
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human