Schlenk tubes, KIMBLE®
Supplier: DWK Life Sciences
Certificates
About this item
This Schlenk glass tube is designed for type 216050 manifolds and performs well under both thermal and pressure-type reactions.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Frequently Bought Together