Magnesium carbonate hydroxide pentahydrate (40.0 - 45.0% MgO) for biochemistry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56378-72-4
:39409-82-0
Formula:C₄H₂Mg₅O₁₄·5H₂O
MDL Number:MFCD00149784
:485.65 g/mol (395.58 anhy)
Melting Pt:350 °C (dec.)
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Titration Complexometric:40.0 to 43.5 % (MgO)
- Arsenic (As):≤4 ppm
- Iron (Fe):≤0.02 %
- Lead (Pb):≤10 ppm
- Acid insoluble substances:≤0.05 % (in HCl)
Specification Test Results
Frequently Bought Together