Order Entry
Slovenia
ContactUsLinkComponent
 
 
 
Sodium nitrate 99+% for biochemistry
 
undefined
 
 
Sodium nitrate 99+% for biochemistry

 

  • Appearance:
    White Crystalline powder
  • pH:
    5.5 to 8.3
  • Titration with HClO4:
    ≥99.0 %
  • Chloride (Cl):
    ≤0.001 %
  • Iodate (IO3):
    ≤0.0005 %
  • Nitrite (NO2):
    ≤0.001 %
  • Phosphate (PO4):
    ≤0.0005 %
  • Sulfate (SO4):
    ≤0.003 %
  • Calcium (Ca):
    ≤0.005 %
  • Iron (Fe):
    ≤0.0003 %
  • Magnesium (Mg):
    ≤0.002 %
  • Heavy metals (as Pb):
    ≤5 ppm
  • Insoluble matter:
    ≤0.005 %

 

 

Frequently Bought Together