
4-Chlorophenylboronic acid pinacol ester 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:195062-61-4
:
Formula:C₁₂H₁₆BClO₂
MDL Number:MFCD05663875
:238.52 g/mol
Melting Pt:55…56 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Solid or crystalline solid
- Infrared spectrum:Conforms
- Melting point:54 to 59 °C
- GC:≥96.0 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
Frequently Bought Together