
6-Quinolineboronic acid pinacol ester 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:406463-06-7
:
Formula:C₁₅H₁₈BNO₂
MDL Number:MFCD06411327
:255.1199 g/mol
Melting Pt:65…67 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to light orange Powder or crystalline solid
- Infrared spectrum:Conforms
- Melting point:62 to 70 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together