D-Biotin-N-succinimidyl ester 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:35013-72-0
:
Formula:C₁₄H₁₉N₃O₅S
MDL Number:MFCD00078531
:341.38 g/mol
Melting Pt:202.3…202.7 °C
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Water:≤2 %
- Elemental Analysis:48.03 to 50.49 % C
- Specific optical rotation:+52° to +54° (20°C, 589 nm) (c=1 in DMF)
Specification Test Results
Frequently Bought Together