
3-Fluoro-4-iodopyridine 97%
Catalog # ACRO436380050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Cream to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:87 to 91 °C
- GC:≥96.0 %
Specification Test Results
About this item
:
:222.99 g/mol
Melting Pt:189.4…190.4 °C
ADR:8,III
:GHS07
Storage Temperature:Ambient
CAS Number:22282-75-3
Formula:C₅H₃FIN
MDL Number:MFCD03092927