
Diisopropylaminomethylidene(diisopropyl)ammonium tetrafluoroborate 97%
Catalog # ACRO435700010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to beige Powder or solid
- Infrared spectrum:Conforms
- NMR:≥96 %
- GC:≥96.0 %
Specification Test Results
About this item
:
:300.19 g/mol
Melting Pt:280…284 °C
:GHS07
Storage Temperature:Ambient
CAS Number:369405-27-6
Formula:C₁₃H₂₉BF₄N₂
MDL Number:MFCD15144860