Order Entry
Slovenia
ContactUsLinkComponent
1-Chlorobutane, anhydrous 99.5%, AcroSeal®
1-Chlorobutane, anhydrous 99.5%, AcroSeal®
Certificates
undefined
1-Chlorobutane, anhydrous 99.5%, AcroSeal®

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.002 %
  • GC:
    ≥99.4 %
  • Residue after evaporation:
    ≤0.001 %

Specification Test Results

Frequently Bought Together