



[3-(4-Morpholinylmethyl)phenyl]magnesium bromide 0.25 M in THF, AcroSeal®
Catalog # 431760500.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:50 mL
- Seq:2370
Specifications
Specification Test Results
- Appearance:Clear yellow Solution
- Titration Grignard:0.15 to 0.35 M
- Refractive index:1.5630 to 1.5650 (20°C, 589 nm)
- GC:≥98.0 %
Specification Test Results
About this item
:
Density:0.926 g/cm³
Flash Pt:–17 °C
ADR:3,II
:280.448 g/mol
Storage Temperature:Ambient
UN:2924
:GHS07
:GHS05
:GHS02
:GHS08
CAS Number:480424-76-8
Formula:C₁₁H₁₄BrMgNO
MDL Number:MFCD03840889