



[2-(4-Morpholinylmethyl)phenyl]magnesium bromide 0.25 M in THF, AcroSeal®
Catalog # 431750500.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:50 mL
- Seq:2370
Specifications
Specification Test Results
- Appearance:Clear yellow Solution
- Titration Grignard:0.15 to 0.35 M
- Titration with NaOH:≥98.5 %
- Chloride (Cl):≤5 ppm
- Phosphate (PO4):≤5 ppm
- Calcium (Ca):≤10 ppm
- Copper (Cu):≤5 ppm
- Iron (Fe):≤5 ppm
- Potassium (K):≤10 ppm
- Sodium (Na):≤20 ppm
- Heavy metals (as Pb):≤5 ppm
- Insoluble matter:≤0.005 % (in water)
Specification Test Results
About this item
:
:280.448 g/mol
Density:0.926 g/cm³
Flash Pt:–17 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
:GHS07
:GHS05
:GHS02
:GHS08
CAS Number:480424-77-9
Formula:C₁₁H₁₄BrMgNO
MDL Number:MFCD03840888