
3,6-Dihydrothiopyran-4-boronic acid pinacol ester
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:862129-81-5
:
Formula:C₁₁H₁₉BO₂S
MDL Number:MFCD10700154
:226.141 g/mol
Melting Pt:42…44.5 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Orange to brown Low melting solid
- Infrared spectrum:Conforms
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together