3,5-Dihydroxybenzaldehyde 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:26153-38-8
:
Formula:C₇H₆O₃
MDL Number:MFCD00016611
:138.12 g/mol
Melting Pt:153…158 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to grey to tan Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤2 %
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together