
Methyl-5-hexynoate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:77758-51-1
:
Formula:C₇H₁₀O₂
MDL Number:MFCD00671366
:126.16 g/mol
Boiling Pt:60…62 °C (10 mmHg)
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4330 to 1.4350 (20°C, 589 nm)
- GC:≥94 %
Specification Test Results
Frequently Bought Together