
Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate 98%
:5909-24-0
:
:C₈H₉ClN₂O₂S
:MFCD00006085
:232.69 g/mol
:60…63 °C
:GHS07
:Ambient
- Appearance:White to yellow Powder
- Infrared spectrum:Conforms
- Melting point:59 to 64 °C
- GC:≥97.5 %
Frequently Bought Together