
3-Hydroxyphthalic anhydride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:37418-88-5
:
Formula:C₈H₄O₄
MDL Number:MFCD00011557
:164.1150 g/mol
Melting Pt:199 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to beige Powder or crystals or flakes
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together