3-Isocyanatopropyltriethoxysilane 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: ICPTES, Triethoxy(3-isocyanatopropyl)silane, (3-Isocyanatopropyl)triethoxysilane, Isocyanic acid 3-(Triethoxysilyl)propyl ester, 3-(Triethoxysilyl)propyl Isocyanate
CAS Number:24801-88-5
:
Formula:C₁₀H₂₁NO₄Si
MDL Number:MFCD00051459
Boiling Pt:130 °C (20 mmHg)
Density:0.99 g/cm³
Molecular Weight:247.366 g/mol
Flash Pt:77 °C
UN:2927
ADR:6.1 II
:GHS06
:GHS08
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4190 to 1.4240 (20°C, 589 nm)
- GC:≥94 %
Specification Test Results
Frequently Bought Together