Palladium hydroxide (20% Pd) on carbon (Pearlman's catalyst) wetted with ca. 60% water, powder, unreduced
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12135-22-7
:
Formula:H₂O₂Pd
MDL Number:MFCD00064599
:140.435 g/mol
Storage Temperature:Ambient
Specification Test Results
- Appearance:Black Powder
- Water:45.0 to 55.0 %
- Palladium (Pd):18.0 to 22.0 % On dry basis
Specification Test Results
Frequently Bought Together