Order Entry
Slovenia
ContactUsLinkComponent
Polyethylene oxide, M.W. 100000-200000
Polyethylene oxide, M.W. 100000-200000
Certificates
undefined
Polyethylene oxide, M.W. 100000-200000

Specification Test Results

  • Appearance:
    White to light yellow Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤2 % (10 g, 50°C, 5 h, 10 mmHg)
  • Viscosity:
    80 to 200 mPa.s (10 % w/w in water at 25°C)

Specification Test Results

Related Information

Frequently Bought Together