Ethyl (R)-(-)-3-hydroxybutyrate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:24915-95-5
:
Formula:C₆H₁₂O₃
MDL Number:MFCD00075386
:132.16 g/mol
Boiling Pt:75…76 °C (12 mmHg)
Density:1.017 g/cm³
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4195 to 1.4215 (20°C, 589 nm)
- GC:≥97.5 %
- Specific optical rotation:-43° to -47° (20°C, 589 nm) (c=1, CHCl3)
Specification Test Results
Frequently Bought Together