Order Entry
Slovenia
ContactUsLinkComponent
D-erythro-Sphingosine (synthetic) 99+%
D-erythro-Sphingosine (synthetic) 99+%
Catalog # ACRO331530500
CAS Number:  
undefined
D-erythro-Sphingosine (synthetic) 99+%
Catalog # ACRO331530500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Appearance:
    White to cream Powder or waxy solid
  • Infrared spectrum:
    Conforms
  • TLC:
    ≥99.0 %

Specification Test Results

About this item

 :
Melting Pt:81…82 °C
 :299.50 g/mol
Storage Temperature:Freezer
CAS Number:123-78-4
Formula:C₁₈H₃₇NO₂
MDL Number:MFCD00036751