
2,5-Pyrazinedicarboxylic acid dihydrate 97%
Catalog # ACRO292320010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to light beige Powder
- Infrared spectrum:Conforms
- Water:16 to 19 % (K.F.)
- Titration with NaOH:≥96.0 %
Specification Test Results
About this item
:
:186.123 g/mol
Melting Pt:257 °C (dec.)
:GHS07
Storage Temperature:Ambient
CAS Number:205692-63-3
Formula:C₆H₆N₂O₅
MDL Number:MFCD00216919