Order Entry
Slovenia
ContactUsLinkComponent
cis-2-Hydroxymethyl-1-cyclohexylamine hydrochloride 99%
cis-2-Hydroxymethyl-1-cyclohexylamine hydrochloride 99%
Catalog # ACRO265860010
CAS Number:  
undefined
cis-2-Hydroxymethyl-1-cyclohexylamine hydrochloride 99%
Catalog # ACRO265860010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to light beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    131 to 135 °C
  • Titration Argentometric:
    ≥98.5 %

Specification Test Results

About this item

 :
 :165.661 g/mol
Melting Pt:131…135 °C
Storage Temperature:Ambient
CAS Number:5691-37-2
Formula:C₇H₁₆ClNO