L-Buthionine sulphoximine 99%
:83730-53-4
:
:C₈H₁₈N₂O₃S
:MFCD00067000
:222.303 g/mol
:234…235 °C (dec.)
:GHS07
:Refrigerator
- Appearance:White Powder
- Infrared spectrum:Conforms
- TLC:≥98.5 %
- Specific optical rotation:+31° to +35° (20°C, 589 nm) (c=1, 1 N HCl)
Frequently Bought Together