(2R,3R,4S,5R)-2-(6-Chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5399-87-1
:
Formula:C₁₀H₁₁ClN₄O₄
MDL Number:MFCD00005738
:286.67 g/mol
Melting Pt:161…163 °C (dec.)
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥99.0 %
Specification Test Results
Frequently Bought Together