Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cefoxitin sodium
Cefoxitin sodium 92.7-97%
Cefoxitin sodium 92.7-97%
Cefoxitin sodium 92.7-97%
J66800.03
:
Thermo Fisher Scientific
:
Cefoxitin sodium 92.7-97%
J66800.03
:
Thermo Fisher Scientific
:
Pk:
1 g
:
:
449.428 g/mol
:
14,1936
:
843 °C
:
>160 °C
:
>110 °C (230 °F)
:
251-574-6
:
GHS07
:
33564-30-6
:
C₁₆H₁₆N₃NaO₇S₂
:
MFCD00079042