DNA sodium salt (Deoxyribonucleic acid sodium salt) (from salmon testes)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Thymonucleic acid sodium salt, DNA sodium salt
CAS Number:9007-49-2
:438545-06-3
MDL Number:MFCD00284534
Merck Index:14,2907
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together