2X YT Microbial medium
J63227.AP
:
- Pk:500 ml
- Description:YT Microbial Medium, 2X
- Environmentally Preferable:
Physical Form: Liquid
Appearance: Yellow-brown
MDL No.: MFCD01636190
Applications: Growth medium for culturing Escherichia coli, particularly laboratory or recombinant strains.