
5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:253870-02-9
:
Formula:C8H9NO3
MDL Number:MFCD06202342
:167.16 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together