Fluorescein disodium salt hydrate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:518-47-8
:
Formula:C₂₀H₁₀Na₂O₅·xH₂O
MDL Number:MFCD00167039
:376.28 g/mol (anhy)
Merck Index:14,4159
Melting Pt:>250 °C
Density:1.601 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-253-0
Related Information
Frequently Bought Together