
4-(Methoxycarbonyl)cyclohexanecarboxylic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:32529-79-6
:
Formula:C₉H₁₄O₄
MDL Number:MFCD06797561
:186.210 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together