

3-Fluoro-4-nitrobenzyl bromide 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:131858-37-2
:
Formula:C₇H₅BrFNO₂
MDL Number:MFCD03094237
:234.024 g/mol
Melting Pt:46…48 °C
ADR:8,II
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together