Triethylene glycol bis(p-toluenesulphonate) 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:19249-03-7
:
Formula:C₂₀H₂₆O₈S₂
MDL Number:MFCD00048096
Melting Pt:80…82 °C
:458.54 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together