L-Phenylalanine ethyl ester hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3182-93-2
:
Formula:C₁₁H₁₆ClNO₂
MDL Number:MFCD00012507
:229.704 g/mol
Melting Pt:153…157 °C
Storage Temperature:Ambient
EINECS:221-673-9
Related Information
Frequently Bought Together