

Lead(II) citrate trihydrate 99%
Catalog # A10701.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:100 g
Specifications
About this item
CAS Number:6107-83-1
:
Formula:C₁₂H₁₆O₁₇Pb₃
MDL Number:MFCD00150456
:1053.843 g/mol
EINECS:208-141-1
ADR:6.1,III
:GHS08
:GHS07
Storage Temperature:Ambient