Poly(methyl methacrylate)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Methacrylic Acid Methyl Ester Polymer, Methyl Methacrylate Polymer, PMMA
CAS Number:9011-14-7
:
Formula:C₅H₈O₂([-CH₂C(CH₃)(CO₂CH₃)-]n)
MDL Number:MFCD00134349
Melting Pt:>150 °C
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together