
Calcium molybdate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7789-82-4
:
Formula:CaMoO₄
MDL Number:MFCD00015976
:200.024 g/mol
Merck Index:14,1681
Melting Pt:965 °C
Density:4.45
EINECS:232-192-9
:GHS07
Storage Temperature:Ambient
Frequently Bought Together