Total Organic Carbon (TOC), standard solution, Specpure®, 1000 µg/ml
Supplier: Thermo Fisher Scientific
Certificates
About this item
Standard for TOC based on potassium hydrogen phthalate (KHC₈H₄O₄).
Physical Form: Liquid
MDL No.: MFCD00133992
Specifications
- MDL No.:MFCD00133992
- Description:Total Organic Carbon (TOC), standard solution
- Physical form:Liquid
Specifications
Related Information
Frequently Bought Together