
Rhodium(II) octanoate dimer
Catalog # 39825.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:778.63 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:73482-96-9
Formula:C₃₂H₆₀O₈Rh₂
MDL Number:MFCD00064724