Anti-Q9VAJ2 Rabbit Polyclonal Antibody
Catalog # ABGEAP20606A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen symbol:Q9VAJ2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Fish
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:43490
- Storage buffer:pH 7,4
- Molecular weight:37.415 kDa
- Sequence:0
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Shipping temperature:4 °C
- Immunogen:This BUB3 antibody is generated from a rabbit immunized with a KLH conjugated synthetic peptide between amino acids from the human region of human BUB3.
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: Q9VAJ2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Fish