Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-SOHLH2 Rabbit Polyclonal Antibody
Catalog # ABGEAP18515C
Supplier:  Abgent
Anti-SOHLH2 Rabbit Polyclonal Antibody
Catalog # ABGEAP18515C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    SOHLH2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    internal
  • Gene ID:
    100526761;54937
  • Amino acid number:
    232 to 258
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    46.941 kDa
  • Sequence:
    DYVKYIREKISPC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.55
  • Shipping temperature:
    4 °C
  • Immunogen:
    This SOHLH2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 232-258 amino acids from the Central region of human SOHLH2.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: SOHLH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human