Anti-HA Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 16B12]
Catalog # AGRIAS152921
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:HA
- Clonality:Monoclonal
- Clone:16B12
- Conjugation:AP (Alkaline Phosphatase)
- Host:Mouse
- Western blot:Yes
- Immunogen:HA-tag: CYPYDVPDYASL
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Mouse anti-HA is a primary antibody which binds to HA and is directly conjugated to Alkaline Phosphatase . This is of advantage to shorten assay time by no need to use a secondary antibody to HA. ,Mouse anti-HA is a primary antibody which binds to HA and is directly conjugated to Alkaline Phosphatase . This is of advantage to shorten assay time by no need to use a secondary antibody to HA.
Type: Primary
Antigen: HA
Clonality: Monoclonal
Clone: 16B12
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype:
Reactivity: