Order Entry
Slovenia
ContactUsLinkComponent
VWR®CryoPro®, Cryogenic storage tanks
VWR®CryoPro®, Cryogenic storage tanks
Catalog # VWRU55709-218
Supplier:  VWR Collection
undefined
VWR®CryoPro®, Cryogenic storage tanks
Catalog # VWRU55709-218
Supplier:  VWR Collection

Specifications

  • Capacity:
    20,5 L
  • Description:
    Canister storaget tank, CC-2, Cryopro®
  • Seq:
    01
  • Model:
    CC-2
  • Weight, full of LN₂:
    28,3 kg
  • Ø×H:
    368×652 mm
  • Weight, empty:
    11,8 kg
  • Ø neck:
    55 mm
  • Pk:
    1

Specifications

About this item

For small to medium capacity applications, the CC series offers traditional canister-style cryogenic storage in efficient narrow neck designs.

CryoRetriever claw-style tool is specifically designed for retrieving boxes, vials, and frames from cryopreservation tanks. Exclusive sealed center shaft prevents hazardous spurting of LN₂ that can occur with a hollow shaft retrieval claw. Aluminium canes simplify and organise cryogenic vials for storage in LN₂ dewars with canisters. One cane holds six 1,2 ml to 2 ml vials.

Sturdy five-wheel rollerbases are available for safely moving full tanks. A low liquid level alarm helps protect against accidental warmups. The phase separator can be attached to the stainless steel transfer hose for increased efficiency and safety when transferring liquid nitrogen.

Caution: All tanks includes canisters.